Brinkmann® Dual Column Heaters, Metrohm
Supplier: Metrohm USA
BMO-LSCH430240 BMO-LSCH460240
21506-152EA 5766.47 USD
21506-152 21506-154
Brinkmann® Dual Column Heaters, Metrohm
Heaters Column Heaters
Column heaters control the temperature of the analysis, by ensuring accurate and reproducible sample results. These both models are designed to work in conjunction with the TC-45 for chromatography application needs "above ambient" range. Wide selection of heaters and controllers designed to be combined for specific needs.
  • Thermostat limits the heater temperature, preventing overheating
  • Pre-heater maximizes column efficiency
  • Two-part aluminum oven with heating element
Ideal when working with a single HPLC column, several extra-long GPC columns, orMore Product Information