Cleanert® CN SPE Plates, 96-Well, Bonna Agela
Supplier: Agela Technologies
SA1002-W SA0502-W SI1002-W SI0502-W
97017-842EA 1932.36 USD
97017-842 97017-844 97017-850 97017-852
Cleanert® CN SPE Plates, 96-Well, Bonna Agela
Microplates Solid Phase Extraction Plates (SPE)
These 96-well plates have a particle size of 40 to 60 µm and a pore size of 60 Å.