Human Peripheral Blood CD8+ T Cells (Cytotoxic T Cells), iXCells Biotechnologies
Supplier: IXCELLS BIOTECHNOLOGIES USA MS
10HU-024-5M
10HU-024-10M
103521-818EA
870.15
USD
103521-818
103521-900
Human Peripheral Blood CD8+ T Cells (Cytotoxic T Cells), iXCells Biotechnologies
Cells
Human Peripheral Blood CD8+ T Cells (Cytotoxic T Cells); Frozen
- High-quality Human Peripheral Blood CD8+ Cells (Helper T Cells)
- High purity
- IRB-approved
Human Peripheral Blood CD8+ Cytotoxic T Cells are isolated from human peripheral blood mononuclear cells (PBMCs) using positive immunomagnetic selection. > 93% of the cells are CD8 positive.