Human Peripheral Blood CD14+ Monocytes, iXCells Biotechnologies
Supplier: IXCELLS BIOTECHNOLOGIES USA MS
10HU-008-10M 10HU-008-40M
103521-806EA 1207.59 USD
103521-806 103521-810
Human Peripheral Blood CD14+ Monocytes, iXCells Biotechnologies
Cells
Human Peripheral Blood CD14+ Monocytes; Frozen
  • High-quality Human Peripheral Blood CD14+ Monocytes
  • High purity
  • IRB-approved
Peripheral Blood CD14+ Monocytes are isolated from normal human peripheral blood mononuclear cells (PBMCs) using positive immunomagnetic selection. > 92% of the cells are CD14+ as showed by flow cytometric analysis