Chloro(1,5-cyclooctadiene)(pentamethylcyclopentadienyl)ruthenium(II) ≥98% for use as a catalyst

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Pentamethylcyclopentadienyl(1,5-cyclooctadiene)ruthenium(II) chloride, RuCl(cod)Cp*

44-0113-250MG 44-0113-1G
100207-680EA 174.09 USD
100207-680 100200-682
Chloro(1,5-cyclooctadiene)(pentamethylcyclopentadienyl)ruthenium(II) ≥98% for use as a catalyst
Chloro(1,5-cyclooctadiene)(pentamethylcyclopentadienyl)ruthenium(II)
Formula: C₁₈H₂₇ClRu
MW: 379.94 g/mol
MDL Number: MFCD07369035
CAS Number: 92390-26-6

Order Now

Specification Test Results

Color and Form Brown Microxtls
Stability Store cold
Purity 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR