D-(+)-Raffinose pentahydrate ≥98%, white crystalline powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: D-(+)-Raffinose pentahydrate, Gossypose pentahydrate, (3R,4S,5S,6R)-2-(((2S,3S,4S,5R)-3,4-Dihydroxy-2,5-bis(hydroxymethyl)tetrahydrofuran-2-yl)oxy)-6-((((3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-3,4,5-triol pentahydrate, Melitriose pentahydrate, O-α-D-Galactopyranosyl-(1→6)-α-D-glucopyranosyl β-D-fructofuranoside, Melitose pentahydrate

114010022 114010032
76019-968EA 764.96 USD
76019-968 76019-970
D-(+)-Raffinose pentahydrate ≥98%, white crystalline powder
D-(+)-Raffinose pentahydrate

Raffinose is a trisaccharide composed of galactose, fructose, and glucose that is used as carbon source in yeast culture media.


  • Suitable for Cell Culture
  • Presentation: White Powder
  • Detection Method: FTIR
  • Soluble in water (50 mg/mL)
  • Store at Room Temperature (15-30 °C)

Formula: C₁₈H₃₂O₁₆·5H₂O
MW: 594.52 g/mol
Melting Pt: 76…86 °C
Density: 1.465 g/mL (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00071590
CAS Number: 17629-30-0

Order Now

Specification Test Results

Identity Test Passes
Purity ≥98%
FTIR Conforms to Standard
Optical Rotation (c=4, water) 101 - 107°
Loss on Drying 13-17%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR