N-(2-Hydroxyethyl)maleimide ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

07-1010-1G 07-1010-250MG
101442-392EA 549.17 USD
101442-392 101442-390
N-(2-Hydroxyethyl)maleimide ≥99%
N-(2-Hydroxyethyl)maleimide
Formula: C₆H₇NO₃
MDL Number: MFCD00465266
CAS Number: 1585-90-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR