Membrane Filter, Mixed Cellulose Esters, Millipore
Supplier: MilliporeSigma
GSWP01300 GSWP03700 GSWP09000 GSWP04700 GSWP14250
220008-144PK 184.62 USD
220008-144 220007-612 220007-220 220007-386 220007-498
Membrane Filter, Mixed Cellulose Esters, Millipore
Filters Membrane Filters
These membrane filter is composed of mixed cellulose membrane and has a 0.22 µm pore size.
  • Biologically inert mixtures of cellulose acetate and cellulose nitrate
  • Hydrophilic
  • White