IPTG (isopropyl-β-D-thiogalactopyranoside) enzyme substrate dioxane-free

Synonyms: IPTG, (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(isopropylthio)tetrahydro-2H-pyran-3,4,5-triol, Isopropyl Beta-D-1-thiogalactopyranoside

R0392 R0393
101673-864EA 321.54 USD
101673-864 101673-866
IPTG (isopropyl-β-D-thiogalactopyranoside) enzyme substrate dioxane-free
IPTG (isopropyl-β-D-thiogalactopyranoside)

The IPTG (isopropyl-beta-D-thiogalactopyranoside) is a highly stable synthetic analog of lactose


It inactivates the lac repressor and induces synthesis of beta-galactosidase, an enzyme that promotes lactose utilization. IPTG is used to induce expression of cloned genes under control of the lac operon It is used in conjunction with X-Gal to determine the lac phenotype in blue or white colony screening.

Formula: C₉H₁₈O₅S
MW: 238.31 g/mol
MDL Number: MFCD00063273
CAS Number: 367-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR