Fermentas Oligo(dT) PCR Primer, Thermo Scientific
SO132 SO131
101674-196EA 220 USD
101674-196 101674-194
Fermentas Oligo(dT) PCR Primer, Thermo Scientific
Nucleic Acid Reagents Primers and Probes
Oligo(dT)18 primer is a synthetic single-stranded 18-mer oligonucleotide with 5'- and 3'-hydroxyl ends.
The primer is supplied as a ready to use, 20X concentrated aqueous solution. Oligo(dT)18 primer has been functionally tested in first strand cDNA synthesis.