Fermentas Maxima™ First Strand cDNA Synthesis Kit, Thermo Scientific
K1642 K1641
101672-962EA 1655.38 USD
101672-962 101672-960
Fermentas Maxima™ First Strand cDNA Synthesis Kit, Thermo Scientific
Nucleic Acid Reagents RT-PCR and cDNA Reagents
The Maxima™ First Strand cDNA Synthesis Kit is a convenient system optimised for use as a first step in two step RT-qPCR. The kit includes the Maxima™ Reverse Transcriptase, which is an advanced enzyme derived by in vitro evolution of M-MuLV RT for RT-qPCR application. The enzyme features high thermostability, robustness and increased synthesis rates in RT-qPCR.
  • High yields of full length cDNA up to 20 kb
  • Efficient cDNA synthesis at wide temperature range from 42 to 65 °C
  • Increased synthesis rate and completion of cDNA synthesis in 15 – 30 minutess
  • High sensitivity and specificity
The Maxima™ First Strand cDNA Synthesis Kit is capable of cDNA synthesis at elevated temperatures (50 to 60 °C) and completion of the reverse transcription reaction in 15 minutes.
In addition, the kit offers highly convenient composition, where Maxima™ Reverse Transcriptase and RiboLock™ RNase Inhibitor are premixed, while all other reaction components: reaction buffer, dNTPs and primers (random hexamers and Oligo(dT)18) are premixed in another tube as a 5X reaction mix.
Información de suministro: Kit includes Maxima™ enzyme mix, 5X reaction mix, nuclease-free water, and detailed protocol.
Precaución: Storage Temperature −20 °C