1-Naphthyl phosphate monosodium salt monohydrate

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

0219941010 0219941001 0219941005
76177-288EA 256.39 USD
76177-288 76177-526 76177-528
1-Naphthyl phosphate monosodium salt monohydrate
1-Naphthyl phosphate monosodium salt monohydrate

A non-specific phosphatase inhibitor. Inhibits acid, alkaline and protein phosphatases.
Preferred substrate for bioanalytical determinations of acid phosphatases, such as prostatic acid phosphatase.
Non-specific phosphatase inhibitor. Inhibits acid, alkaline and protein phosphatases.
+4°C


  • Presentation: White Powder
  • Solubility: H2O: soluble (50 mg/mL )

Formula: C₁₀H₈NaO₄P·1H₂O
Storage Temperature: Refrigerator
MDL Number: MFCD00150615
CAS Number: 81012-89-7

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR