Uridine-5′-diphosphogalactose disodium salt

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: Udp-alpha-d-galactose disodium salt, UDP-galactose, Uridine-5'-diphospho-a-D-galactose disodium salt, UDP-Ga

0219590505 0219590510 0219590525
76177-472EA 89.7 USD
76177-472 76177-474 76177-476
Uridine-5′-diphosphogalactose disodium salt
Uridine-5′-diphosphogalactose disodium salt

Uridine-5'-diphosphogalactose Disodium Salt is a donor substrate for galactosyltransferases involved in the addition of galatose (galactosylation) molecules to N-linked and O-linked oligosaccharides and glycerolipids (biosynthesis of galactose-containing oligosaccharides).
UDP-Gal and its analogues are used to study the distribution, specificity and kinetics of galactosyltransferase(s).
Store at -0 °C, desiccate.


  • Presentation: Powder
  • Soluble in water (100 mg/mL-clear colorless solution).

Formula: C₁₅H₂₃N₂Na₂O₁₇P₂
MW: 611.27 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00077895
CAS Number: 137868-52-1

Order Now

Specification Test Results

Identity Test Passes
Purity ≥95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR