L(-)-Carnitine hydrochloride

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: Levocarnitine hydrochloride, L-Carnitine hydrochloride, (R)-3-Carboxy-2-hydroxy-N,N,N-trimethylpropan-1-aminium chloride, LC-80, (R)-3-Hydroxy-4-(trimethylammonio)butyrate hydrochloride, L-Carnitine chloride, (R)-Carnitine hydrochloride, Levocarnitine chloride

0219503101 0219503105 0219503125
76177-260EA 48.27 USD
76177-260 76177-262 76177-410
L(-)-Carnitine hydrochloride
L(-)-Carnitine hydrochloride

Carnitine is a quaternary amine that occurs naturally in most mammalian tissue.
It is present in relatively high concentrations in skeletal muscle and heart where it is involved in regulating energy metabolism. It shifts glucose metabolism from glycolysis to glycogen storage and enhances the transport of long chain fatty acids into the mitochondria where they are oxidized for energy production.
Store at Room Temperature(15-30 °C), desiccate


  • Presentation: White Crystalline Powder
  • Soluble in water; insoluble in acetone.

Formula: C₇H₁₆ClNO₃
MW: 197.66 g/mol
Melting Pt: 142 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00066100
CAS Number: 6645-46-1

Order Now

Specification Test Results

Identity Test Passes
Purity ~98%
Optical Rotation -20° to -25° (c=1, water)
FTIR Conforms to Standard
Residue on Ignition ≤0.5%
Heavy Metals ≤10ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR