Sodium dodecylbenzenesulfonate ≥80%, off-white with orangish hue flakes

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: Laurylbenzenesulphonic acid Sodium Salt, Alkylbenzenesulphonic acid Sodium Salt, Sodium Alkylbenzenesulphonate, Dodecylbenzenesulphonic acid sodium salt, Sodium Laurylbenzenesulphonate

0215788983 0215788991
76102-760EA 64.16 USD
76102-760 76102-762
Sodium dodecylbenzenesulfonate ≥80%, off-white with orangish hue flakes
Sodium dodecylbenzenesulphonate

Dodecylbenzene Sulfonic Acid is a member of the linear alkylbenzenesulfonates.


  • Off-white fakes with orangish hue


Sodium dodecylbenzenesulfonate was used to fabricate amperometic biosensors for glucose.
Sodium dodecylbenzenesulfonate (SDBS) acts as a surfactant in the ionic self-assembly reaction of cyclophane BIMCP-1.


Soluble in 10% aqueous clear, slightly yellow solution.

Formula: C₁₈H₂₉NaO₃S
MW: 348.48 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011508
CAS Number: 25155-30-0
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceb901522021425d8ec257bf4f9ad7e99ab67a30&vid=dcaabaace5cd726641495dbd5082bc58b246c528&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity ≥80%
FTIR Conforms to Reference

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR