Phosphotungstic acid hydrate, white crystalline powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Danger
|
Phosphotungstic acid hydrate is a heteropoly acid.
Phosphotungstic acid is used in histology as a component for staining of cell specimens. It is a common negative stain for viruses, nerves, polysaccharides, and other biological tissue materials for imaging by a transmission electron microscope. Phosphotungstic acid has been used to precipitate different types of dyes as lakes.
Store at Room Temperature(15-30 °C)
- Presentation: White Crystalline Powder
- Soluble in water(1 g + 10 mL, H2O), alcohol and ether.
Formula:
H₃[P(W₃O₁₀)₄]·nH₂O Melting Pt: ~95 °C (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00011342 CAS Number: 12501-23-4 |
|
Specification Test Results
Identity Test | Passes |
Sodium (Na) | ≤0.01% |
Iron (Fe) | ≤0.003% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...