Flavin adenine dinucleotide disodium salt ≥95%, orange powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: (((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl)methoxy)(((((2R,3S,4S)-5-(7,8-dimethyl-2,4-dioxo-2H,3H,4H,10H-benzo[g]pteridin-10-yl)-2,3,4-trihydroxypentyl)oxy)(hydroxy)phosphoryl)oxy)phosphinic acid Disodium Salt, FAD-Na2, Flavin Adenine Dinucleotide Disodium Salt, Riboflavin 5'-adenosine diphosphate disodium salt, FAD Disodium Salt

0215112780 0215112783 0215112790
76102-634EA 153.16 USD
76102-634 76102-636 76102-712
Flavin adenine dinucleotide disodium salt ≥95%, orange powder
Flavin adenine dinucleotide disodium salt

Flavin Adenine Dinucleotide is a prosthetic group of certain oxidases.
Flavin adenine dinucleotide (FAD) is used as a redox cofactor (electron carrier) by flavoproteins including succinate dehydrogenase (complex), α-ketoglutarate dehydrogenase, apoptosis-inducing factor 2 (AIF-M2, AMID), folate/FAD-dependent tRNA methyltransferases, and N-hydroxylating flavoprotein monooxygenases. FAD is a component of the pyruvate dehydrogenase complex.
Store at -0 °C.


  • Presentation: Orange Powder
  • Soluble in water: 50 mg/mL.

Formula: C₂₇H₃₃N₉O₁₅P₂
MW: 829.52 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00151217
CAS Number: 84366-81-4

Order Now

Specification Test Results

Identity Test Passes
Purity ≥95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR