Chitosan (from Crustacean chitin), off-white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0215059710 0215059725 0215059780 0215059783
76102-682EA 60.34 USD
76102-682 76102-684 76102-686 76102-688
Chitosan (from Crustacean chitin), off-white powder
Chitosan

Chitosan is a cationic marine polymer derived form the shells of crustaceans (e.g., crabs). MP's chitosan is a partially de-acetylated chitin formed by the reaction of chitin with concentrated alkali. Chitin is a high molecular weight linear polymer of anhydro-N-acetyl-glucosamine. Chitin occurs in nature as part of the protective coating of some invertebrates.


Chitosan forms gels with multivalent anions. Gives clear solutions that dry to strong, clear films. It is biocompatible, antibacterial and environmentally friendly polyelectrolyte with a variety of applications including water treatment, chromatography, additives for cosmetics, textile treatment for antimicrobial activity, novel fibers for textiles, photographic papers, biodegradable films, biomedical devices, and microcapsule implants for controlled release in drug delivery.


Precaución: Research Use Only (RUO).

Density: 0.15…0.3 g/cm³ (bulk density)
Storage Temperature: Ambient
MDL Number: MFCD00161512
CAS Number: 9012-76-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c5c7c95682b35613228f7f320e7460bb1f8ae2b&vid=98f9d70010d3bf7e7c9c1a900c8f68e4335858ab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Source Crustacean Chitin
Moisture ≤10%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR