L-Cystine dihydrochloride, white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
(2R,2'R)-3,3'-Disulphanediylbis(2-aminopropanoic acid) dihydrochloride, H-Cystine-OH·2HCl
0210557605
0210557625
0210557680
76102-656EA
52.74
USD
76102-656
76102-658
76102-660
L-Cystine dihydrochloride, white powder
L-Cystine dihydrochloride
L-Cystine is a natural proteinogenic amino acid sulfide dimer of L-cysteine.
L-Cystine is a natural proteinogenic amino acid sulfide dimer of L-cysteine. L-cystine is used as a cell culture supplement. L-cystine is also used to characterize and identify specific amino acid transport systems in cells such as system system Xc(─) .
Store at Room Temperature(15-30 °C)
- Presentation: White Powder
- Soluble in Water
Formula:
C₆H₁₂N₂O₄S₂·2HCl MW: 313.23 g/mol Melting Pt: 228…232 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00070399 CAS Number: 30925-07-6 |
Specification Test Results
Identity Test | Passes |
FTIR | Conforms to Standard |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...