BSA Gold Tracer, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
25487 25483 25486 25493 25489 25490 25492 25481 25480 25484
100487-912EA 254.71 USD
100487-912 100487-906 100487-910 100487-920 100487-914 100487-916 100487-918 100487-904 100487-902 100487-908
BSA Gold Tracer, Electron Microscopy Sciences
Protein Linking and Labeling Systems
BSA coated gold tracers are useful for detecting open connections and tissue damage.
  • Particle sizes of 0.8, 6, 10, 15 and 25 nm
  • Shelf life of 18 months
  • Prepared with BSA-c™ as particle conjugated protein
Aurion Gold Tracers are used to visualize charged moieties in specimens: anionic tracers bind to polycationic moieties (basic proteins e.g
AURION Gold Tracers are available in 5 and 10 mL volume packages and are supplied at an OD 520nm of 2.0 for the conventional particle size range and at equivalent OD for the Ultra Small tracers.
histones), cationic tracers bind to polyanionic moieties (membranes, acidic proteins). Under proper conditions and with suitable microscopical techniques, the tracers can also be used to follow cellular events in time.
Información de suministro: Supplied in PBS, with 15 mM NaN3.
Precaución: The products should be stored at 4 to 8 °C. Freezing is not recommended. AURION Gold Tracers are shipped containing NaN3 as preservative. If they are intended for use in living organisms, the preservative has to be removed prior to use.