EDTA disodium salt dihydrate ≥99%, white crystalline powder for molecular biology

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Ethylenediaminetetraacetic acid, disodium salt, dihydrate, Ethylenediaminetetraacetic acid disodium salt dihydrate, Kestranal® 2S dihydrate, Disodium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenedinitrilotetra-acetic acid disodium salt dihydrate, Ethylenediaminetetraacetic acid disodium dihydrate, Titriplex III, Ethylenediaminetetra-acetic acid disodium salt dihydrate, Kestranal® 2S 2 H2O, Sequestrene Na₂, Edetate disodium dihydrate, Edetic acid disodium salt dihydrate

0219482250 0219482280 0219482283 0219482290 0219482291
76177-944EA 50.08 USD
76177-944 76177-946 76177-948 76177-950 76177-952
EDTA disodium salt dihydrate ≥99%, white crystalline powder for molecular biology
EDTA disodium salt dihydrate

Storage: Store at Room Temperature (15-30 °C).
Ethylenediamine Tetraacetic Acid is a polyamino carboxylic acid hexadentate ligand and a chelating agent. This product is designated as Molecular Biology grade and is suitable for molecular biology applications.
Ethylenediamine Tetraacetic Acid is a chelator of divalent cations. It also inhibits enzymes, such as metalloproteases, that require divalent cations for activity.


  • Molecular Biology Reagent Grade
  • Presentation: White Crystalline Powder
  • pKa: 2.0, 2.7, 6.2, 10.3 (Lit.)
  • Soluble in water : 0.1 M, clear, colorless aqueous solution.

Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
MW: 372.24 g/mol
Melting Pt: 255
Storage Temperature: Ambient
MDL Number: MFCD00150037
CAS Number: 6381-92-6
Merck Index: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c101ae031578eff4036863ff241d4176bfba0a8f&vid=a4d43887181c38936cedcd8d634289941e3487dc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard
pH (1% aq soln) 4-6
Loss on Drying ≤10%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR