Potassium dihydrogen phosphate, anhydrous, white crystalline powder cell culture reagent

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: Monopotassiumphosphate, prim.-Potassium phosphate, Monopotassium phosphate, Potassium phosphate monobasic, Monobasic Potassium Phosphate, Potassium dihydrogen orthophosphate

0219472780 0219472790 0219472791
76177-918EA 37.39 USD
76177-918 76177-920 76177-922
Potassium dihydrogen phosphate, anhydrous, white crystalline powder cell culture reagent
Potassium dihydrogen phosphate

Storage: Room Temperature, desiccate
Potassium phosphate is a reagent with high buffering capacity. It occurs in several forms: monobasic, dibasic and tribasic (K3PO4). Most pH neutral potassium phosphate buffer solutions consist of mixtures of the monobasic and dibasic forms to varying degrees, depending on the desired pH.
Potassium phosphate monobasic is typically used as a component for a wide variety of media used in the culture of microorganisms. It is widely used in molecular biology, biochemistry, and chromatography. It is a component in phosphate buffered saline (PBS). In addition to helping maintain pH, it supplies essential phosphate.


  • Anhydrous
  • Cell Culture Reagent Grade
  • Presentation: White Crystalline Powder
  • pKa: (1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid) at 25 °C (Lit.)
  • Soluble in water (100 mg/mL); Insoluble to slightly soluble in ethanol; insoluble in alcohol.

Formula: KH₂PO₄
MW: 136.09 g/mol
Melting Pt: 250 °C (dec., lit.)
Density: 2.338 g/mL (25 °C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00011401
CAS Number: 7778-77-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8cc7bd62e6990fd4b85b54f31f6bab3aa5b8494&vid=4dc7a0f7c05b5aeeba29cb5c37e5301b9a520fac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Assay ≥99.0%
pH (5% aq soln) 4.1-4.5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR