L(-)-Glutathione (reduced form) ≥98%, off-white powder cell culture reagent

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: L-Glutathione Reduced, GSH, (S)-2-Amino-5-(((R)-1-((carboxymethyl)amino)-3-mercapto-1-oxopropan-2-yl)amino)-5-oxopentanoic acid, Glutathione Reduced

0219467910 0219467925 0219467980 0219467905
76177-852EA 174.44 USD
76177-852 76177-854 76177-856 76177-906
L(-)-Glutathione (reduced form) ≥98%, off-white powder cell culture reagent
L(-)-Glutathione (reduced form)

Storage: Store at +4 °C, store under nitrogen
Glutathione is the major low molecular weight thiol compound of the living plant or animal cell. It is a tripeptide with a gamma peptide linkage between the amine group of cysteine (which is attached by normal peptide linkage to a glycine) and the carboxyl group of the glutamate side-chain. It is an antioxidant, preventing damage to important cellular components caused by reactive oxygen species such as free radicals and peroxides. The sulfhydryl (thiol) group (SH) of cysteine serves as a proton donor and is responsible for the biological activity of glutathione.
Glutathione suppresses human immunodeficiency virus expression in chronically infected monocytic cells. It is a useful tripeptide involved in many aspects of metabolism, including transport of g-glutanyl amino acids and reductive cleavage of disulfide bonds.
Endogenous antioxidant that plays a major role in reducing reactive oxygen species formed during cellular metabolism and the respiratory burst. Glutathione-S-transferase catalyzes the formation of glutathione thioethers with xenobiotics, leukotrienes, and other molecules that have an electrophilic center. Glutathione also forms disulfide bonds with cysteine residues in proteins. Via these mechanisms, it can have the paradoxical effect of reducing the efficacy of anti-cancer agents.


  • Cell Culture Reagent Grade
  • Presentation: Off-White Powder
  • Amino Acid Sequence :γ-Glu-Cys-Gly
  • Soluble in water giving clear and colorless solution (3.5% aq soln).

Formula: C₁₀H₁₇N₃O₆S
MW: 307.33 g/mol
Melting Pt: 192…195 °C (lit.)
Storage Temperature: Refrigerator
MDL Number: MFCD00065939
CAS Number: 70-18-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c0ecec88b19c6b78946b004e29823b962632794&vid=db43a6c82bc0c3243287483dc3dc49e522e99659&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test A Passes
Identity Test B Passes
Purity ≥98%
FTIR Conforms to Standard
Solubility (3.5% aq soln) Clear, Colorless Solution

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR