L(+)-Glutamine ≥99%, white powder cell culture reagent

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Gln, L-Glutamine, H-Gln-OH, (S)-(+)-Glutamine, (S)-2,5-Diamino-5-oxopentanoic acid, Levoglutamide, Q

0219467825 0219467883
76177-780EA 39.51 USD
76177-780 76177-900
L(+)-Glutamine ≥99%, white powder cell culture reagent
L(+)-Glutamine

Storage: Store at Room Temperature (15-30 °C)
L-Glutamine, the uncharged and amidated analog of L-glutamic acid, is an important amino acid for the incorporation of NH4+ into biomolecules. It is biosynthesized from NH4.
L-Glutamine is an essential amino acid that is a crucial component of culture media that serves as a major energy source for cells in culture.
L-Glutamine is an essential amino acid that is a crucial component of culture media that serves as a major energy source for cells in culture. L-Glutamine is very stable as a dry powder and as a frozen solution. In liquid media or stock solutions, however, L-glutamine degrades relatively rapidly. Optimal cell performance usually requires supplementation of the media with L-glutamine prior to use.


  • Cell Culture Reagent Grade
  • Presentation: Free Flowing, White Powder
  • Soluble in water (25 mg/mL)

Formula: C₅H₁₀N₂O₃
MW: 146.15 g/mol
Melting Pt: 185 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00008044
CAS Number: 56-85-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8dfea730b88e8a54acd62e1064c2a5bff0853b3f&vid=4afe9fa2191274e8dd4ea12ba1278b66507753b9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99%
Residue on Ignition <0.1%
Loss on Drying <0.5%
Optical Rotation +30 to +35° (c=3.5,1N HCl)
pH (2%, aq soln) 4.5-6.5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR