Iron(II) sulfate heptahydrate ≥99%, bluish green granular powder cell culture reagent

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: Iron vitriol, Ferrous sulphate heptahydrate

0219466391 0219466383
76177-712EA 162.33 USD
76177-712 76177-848
Iron(II) sulfate heptahydrate ≥99%, bluish green granular powder cell culture reagent
Iron(II) sulphate heptahydrate

Storage: Room Temperature, desiccate, store under nitrogen.
Ferrous sulfate heptahydrate is a reagent used in the manufacturing of iron compounds, in electroplating baths, aluminium etching, process engraving and lithography, and in redox polymerisation. It is used as a reducing agent in chemical processes, leather dyes and writing inks. Also, it is a component of weed killers, wood preservatives and water treatment processes. Used In prevention of chlorosis in plants and therapeutically as hematinic. In veterinary medicine, it is used as an astringent to combat iron deficiency. Ferrous sulfate heptahydrate is used in the determination of phosphorus as per the method of Taussky and Shorr. Ferrous sulfate heptahydrate is used in cell culture applications generally bound to transferrin, citrate or other iron transport molecules.


  • Cell Culture Reagent Grade
  • Auto Ignition: > 100 °C (Lit.)
  • Vapor Pressure: 14.6 mmHg ( 25 °C)
  • Presentation: Bluish Green Granular Powder
  • Soluble in water (100 mg/mL).

Formula: FeSO₄·7H₂O
MW: 278.02 g/mol
Boiling Pt: 2735 °C (1013 hPa) (lit.)
Melting Pt: 1539 °C (lit.)
Density: 1.898 g/mL (25 °C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00149719
CAS Number: 7782-63-0
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c9d2eb97ada0c8fdc5b8aed0b2cd61ab7a86347&vid=67a277ecd97dae89778bb11d863cfd9a86c041c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99%
FTIR Conforms to Reference

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR