Polyethylene glycol 8,000, white prills for molecular biology

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

02194839.5 0219483901 0219483902
76177-402EA 83.16 USD
76177-402 76177-404 76177-406
Polyethylene glycol 8,000, white prills for molecular biology
Polyethylene glycol 8,000

Polyethylene glycol (PEG) is a condensation polymer of ethylene oxide and water.
Polyethylene Glycol has been used in the precipitation of proteins, as a fusing agent in enhancing the effect of macrophages on hybridoma, in the separation and purification of biomolecules and in induction of cell hybridization.
PEG's are susceptible to oxidative degradation in the presence of air. Minimizing the exposure of PEG to elevated temperatures and/or exposure to oxygen, or addition of an antioxidant can limit the amount of degradation. PEGs do not hydrolyze or deteriorate upon storage. PEGs do not support the growth of molds.


  • Molecular Biology Reagent Grade
  • Presentation: White Prills
  • Vapor Density: >1 (vs air) (Lit.)
  • Vapor Pressure: <0.01 mmHg at 20 °C (Lit.)
  • Average value of n = 181.4
  • Soluble in water (approximately 630 mg/mL, 20 °C). PEGs are also soluble in many polar solvents such as acetone, alcohols and chlorinated solvents. They are insoluble in nonpolar solvents such as hydrocarbons.

Formula: H(OCH₂CH₂)nOH
Melting Pt: 56…66 °C
Density: 1.0845 g/mL (70 °C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00131611
CAS Number: 8042-47-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a4d3ffdb62509fa565f545135ee35822fa93fc73&vid=4bb7ebcd3030188c4e4e0fa3cb8ba27efe60fcd4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
FTIR Confrorms to Standard
pH (5% aq soln) 5.0-7.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR