Imidazole ≥99%, white crystalline powder for molecular biology

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

0219482905 0219482925 0219482980
76177-220EA 95.98 USD
76177-220 76177-222 76177-224
Imidazole ≥99%, white crystalline powder for molecular biology
Imidazole

Imidazole is a histamine antagonist.
Imidazole can be used to prepare buffers in the pH range of 6.2-7.8 at 25 °C. It is also a chelator for the binding of various divalent cations.
Solutions can be successfully sterilized by autoclaving, and are stable for at least 2 years at 2-8 °C, protected from light


  • Molecular Biology Reagent Grade
  • Vapor Pressure: 0.003 hPa at 20 °C (Lit.)
  • Presentation: White Crystalline Powder
  • pKa: 7.1 (Lit.)
  • Soluble in water (approximately 500 mg/mL), yielding a clear solution.

Formula: C₃H₄N₂
Boiling Pt: 268 °C (lit.)
Melting Pt: 85…95 °C
Density: 500…600 kg/m³ (bulk density)
Storage Temperature: Ambient
MDL Number: MFCD00005183
CAS Number: 288-32-4
UN: 3263
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f193ab88a37d9067d072717016a5939a3e2570d3&vid=9ec2cc640d59f86b903ba299f170c83cb1c1a058&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48eba49e61201d6fffa2bef0c54633a20660c7ef&vid=e7f154e4976b52732539e49bbff60454d5eb82ff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard
Melting Point 85°-95°C
pH (1% aq soln) >7
Water Content ≤0.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR