HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid ≥99%, white powder for molecular biology

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0219482725 0219482780 0219482790
76177-214EA 76.45 USD
76177-214 76177-216 76177-218
HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid ≥99%, white powder for molecular biology
HEPES (2-[4-(2-hydroxyethyl)-1-piperazinyl]ethanesulphonic acid)

HEPES is a zwitterionic organic chemical buffering agent.
HEPES is widely used in many biochemical reactions and as a buffering agent in some cell culture media.


  • Molecular Biology Reagent Grade
  • Presentation: White Powder
  • pKa: 7.5 (25 °C) (Lit.)
  • Solutions may be autoclaved under standard conditions

Formula: C₈H₁₈N₂O₄S
Melting Pt: 234 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00006158
CAS Number: 7365-45-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4cdc09c866676149941b93b2cac8d453175059a2&vid=0b09e5b84ea34a806443c3071fad63e15e4e3433&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=857b891cd5e19daf7ea54a1f490de8c442eeb208&vid=857b891cd5e19daf7ea54a1f490de8c442eeb208&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99%
OD250 (10% aq soln) <0.2
OD260 (10% aq soln) <0.1
OD280 (10% aq soln) <0.1
pH (1% aq soln) 4.0-5.5
Moisture <1.0%
Solubility (10% aq soln) Clear, Colorless Solution


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR