Calcium chloride dihydrate ≥99%, white powder cell culture reagent

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

02194635.5 0219463501
76177-042EA 87.08 USD
76177-042 76177-044
Calcium chloride dihydrate ≥99%, white powder cell culture reagent
Calcium chloride dihydrate

Calcium Chloride is a salt of calcium and chloride. It behaves as a typical ionic halide, and is solid at room temperature.
Calcium chloride is a commonly used reagent in biochemistry. It plays important roles in many biological processes, including signal transduction, muscle contraction, and maintenance of cell membrane and cell wall stability. Calcium chloride is used in the preparation and transformation of competent E. coli and in the transfection of eukaryotic cells with either plasmid DNA or high molecular weight genomic DNA. It has been used in the stabilization and two dimensional crystallization of the NADH-ubiquinone oxidoreductase (complex I) from Escherichia coli. It is also used as a dessicant, coagulant in rubber manufacturing and as an accelerator in the concrete industry.


  • Cell Culture Reagent Grade
  • Presentation: White Powder
  • Soluble in water - 294 mg/mL (clear, colorless solution) with heat liberation, ethanol, acetic acid, acetone.

Formula: CaCl₂·2H₂O
Melting Pt: 176 °C (dec., lit.)
Storage Temperature: Ambient
MDL Number: MFCD00149613
CAS Number: 10035-04-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=012d1dac588a2482dd4270022cdc6f3a0d9cc1ec&vid=c9f5c63a6dd1c97ddfd6fc46a5b392ba3dfeb70b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity ~99%
pH (5% aq soln) 4 - 9.5
Ammonium ≤0.005%
Barium ≤0.005%
Heavy Metals ≤5ppm
Insoluble Matter ≤0.01%
Iron ≤5ppm
Magnesium ≤0.005%
Oxidizing Substances ≤0.003%
Potassium ≤0.01%
Sodium ≤0.02%
Strontium ≤0.1%
Sulfate ≤0.01%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR