Arachidonic acid ≥98%, clear, colorless liquid cell culture reagent

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: (5Z,8Z,11Z,14Z)-Icosa-5,8,11,14-tetraenoic acid

0219462510 0219462550 0219462580 0219462590
76177-034EA 81.2 USD
76177-034 76177-036 76177-038 76177-040
Arachidonic acid ≥98%, clear, colorless liquid cell culture reagent
Arachidonic acid

Arachidonic Acid is an essential fatty acid. Occurs in liver, brain, glandular organs, and depot fats of animals, in small amounts in human depot fats, and is a constituent of animal phosphatides.
Arachidonic Acid is a precursor in the biosynthesis of prostaglandins, thromboxanes, and leukotrienes. Arachidonic acid plays a key role in cellular regulation and is controlled through multiple interconnected pathways.
Arachidonic acid (AA) is an unsaturated ω6 fatty acid constituent of the phospholipids of cell membranes. Phospholipase A2 releases AA from the membrane phospholipids in response to inflammation. AA is subsequently metabolized to prostaglandins and thromboxanes by at least two cyclooxygenase (COX) isoforms, to leukotrienes and lipoxins by lipoxygenases, and to epoxyeicosatrienoic acids via cytochrome p450-catalyzed metabolism. AA and its metabolites play important roles in a variety of biological processes, including signal transduction, smooth muscle contraction, chemotaxis, cell proliferation and differentiation, and apoptosis. AA has been demonstrated to bind to the a subunit of G protein and inhibit the activity of Ras GTPase-activating proteins (GAPs). Cellular uptake of AA is energy dependent and involves protein-facilitated transport across the plasma membrane.
If ethanol is undesirable, arachidonic acid may be dissolved in acetonitrile, DMF, or DMSO. Simply evaporate the ethanol under a gentle stream of nitrogen (be certain not to evaporate the material to dryness) and redissolve the arachidonic acid in the solvent of choice.Just prior to use, make dilutions of the stock solution into aqueous buffer or isotonic saline to bring the arachidonic acid to the desired concentration. Ensure that the residual amount of organic solvent is insignificant, since organic solvents may have physiologic effects at low concentrations. A control using the solvent in the absence of the prostaglandin will address this potential variable. We do not recommend storing the aqueous solution for more than one day. It is difficult to obtain aqueous solutions of arachidonic acid directly. However, an organic solvent free solution of arachidonic acid can be prepared using concentrated basic buffers (pH > 8.0 and ionic strength not less than 0.1 M). Add 400 μL of cold buffer (0 °C) per mg of arachidonic acid and agitate vigorously and/or ultrasonicate.


  • Cell Culture Reagent Grade
  • Presentation: Clear, Colorless Liquid
  • Refractive Index: n20/D 1.4872(Lit.)
  • Soluble in chloroform (50 mg/mL) and absolute ethanol (10 mg/mL) (Lit.)

Formula: C₂₀H₃₂O₂
MW: 304.5 g/mol
Boiling Pt: 170 °C (0.2 mmHg)) (lit.)
Melting Pt: –49 °C (lit.)
Density: 0.92 g/ml
Storage Temperature: Freezer
MDL Number: MFCD00004417
CAS Number: 506-32-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b61646c20fe9c972af2785e4936e96544065b3cb&vid=2df6b8e40ae47efa112611431315ade6bfbd04a7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity ≥98%
Source Fungal

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR