1,2-Dimethoxyethane ≥99% inhibitor-free, Reagent Grade

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Dimethyl glycol, Ethylene glycol dimethyl ether, DME

E27408-1L E27408-2.5L E27408-20L
BJE27408-1LEA 274 USD
BJE27408-1L BJE27408-2.5L BJE27408-20L
1,2-Dimethoxyethane ≥99% inhibitor-free, Reagent Grade
1,2-Dimethoxyethane
Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Boiling Pt: 85 °C (1.013 hPa)
Melting Pt: -58 °C
Density: 0.87 g/cm³ (20 °C)
Flash Pt: -2 °C
Storage Temperature: Ambient
MDL Number: MFCD00008502
CAS Number: 110-71-4
EINECS: 203-794-9
UN: 2252
ADR: 3,II

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 99.0 %
Water (Karl Fischer) Max. 0.5 %
Peroxides (as H₂O₂) Max. 15 ppm
Vapor Pressure 48 mmHg ( 20 °C)
Explosion Limit 10.4 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR