n-Decane 99, reagent

Supplier: Honeywell Research Chemicals
Danger

D901-100ML D901-500ML
BJD901-100MLEA 73.66 USD
BJD901-100ML BJD901-500ML
n-Decane 99, reagent
n-Decane
Formula: H₃C(CH₂)₈CH₃
MW: 142.28 g/mol
Boiling Pt: 174.1
Melting Pt: -30
Density: 0.732 g/cm³ (20 °C)
Flash Pt: 57.4 °C (1.013.25 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00008954
CAS Number: 124-18-5
EINECS: 204-686-4
UN: 2247
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0e4dead3a7841a2258e8a5b99b880895cd06aa1&vid=209e0ebbe6a38426575fcf5608cf6f2b586e6d5e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Vapor Pressure 3.77 mmHg ( 37.7 °C)
Identity (IR) complying
Assay (GC) Min. 99.0 %
Refractive index (n 20/D) 1.409 - 1.413
Explosion Limit 2.6 %
Autoignition Temperature 410 °F
Vapor Density 4.9 (vs air)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR