Decahydronaphthalene (mixture of cis and trans isomers) 98%, Reagent Grade
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
Decalin, Bicyclo[4.4.0]decane, Decahydronaphthalene
D251-20L
D251-4L
BJD251-1LEA
156.07
USD
BJD251-1L
BJD251-20L
BJD251-4L
Decahydronaphthalene (mixture of cis and trans isomers) 98%, Reagent Grade
Decahydronaphthalene (mixture of cis and trans isomers)
Formula:
C₁₀H₁₈ Boiling Pt: 4.9 %(V) Melting Pt: -40 °C (1.013 hPa) Density: 0.88 g/cm³ (20 °C) Flash Pt: 57 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004130 CAS Number: 91-17-8 EINECS: 202-046-9 UN: 1147 ADR: 3,III |
|
Specification Test Results
Vapor Pressure | 42 mmHg ( 92 °C) |
Identity (IR) | complying |
Assay on cis/trans-isomers (GC) | Min. 97.5 % |
Explosion Limit | 0.7-4.9 %, 100 °F |
Refractive Index | n20/D 1.474 (lit.) |
Vapor Density | 4.76 (vs air) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...