Orthophosphoric acid ≥99.0% (by titrimetric analysis), crystallised, Puriss. p.a., Fluka™

Supplier: Honeywell Research Chemicals
Danger

79622-1KG 79622-500G
BJ79622-1KGEA 217 USD
BJ79622-1KG BJ79622-500G
Orthophosphoric acid ≥99.0% (by titrimetric analysis), crystallised, Puriss. p.a., Fluka™
Orthophosphoric acid
Formula: H₃PO₄
Melting Pt: 42 °C
Density: 1.880 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011340
CAS Number: 7664-38-2
UN: 3453
ADR: 8,III

Order Now

Specification Test Results

Assay 99.0 - 101.0 %
Melting point 38 - 42 °C
Arsenic (As) Max. 1 ppm
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 1 ppm
Cobalt (Co) Max. 1 ppm
Chromium (Cr) Max. 1 ppm
Copper (Cu) Max. 1 ppm
Iron (Fe) Max. 10 ppm
Potassium (K) Max. 5 ppm
Magnesium (Mg) Max. 1 ppm
Manganese (Mn) Max. 1 ppm
Sodium (Na) Max. 50 ppm
Nickel (Ni) Max. 1 ppm
Lead (Pb) Max. 1 ppm
Zinc (Zn) Max. 1 ppm
Chloride (Cl) Max. 5 ppm
Nitrate (NO₃) Max. 5 ppm
Sulfate (SO₄) Max. 30 ppm
Phosphite,Hypoph. (H₃PO₃) Max. 500 ppm
KMnO₄ reducing matter (as O) Max. 20 ppm
Vapor Pressure 5 mmHg ( 25 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR