Diethyl ether, anhydrous ≥99.0% contains inhibitor ACS, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger


Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

673811-4L 673811-4X4L
BJ673811-4LEA 751.9 USD
BJ673811-4L BJ673811-4X4L
Diethyl ether, anhydrous ≥99.0% contains inhibitor ACS, Riedel-de Haën™
Diethyl ether

ACS GENERAL USE SOLVENTS


  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications


Beilstein Number: 1696894
RTECS Number: KI5775000
UNSPSC Code: 12191602

Formula: (CH₃CH₂)₂O
MW: 74.12 g/mol
Boiling Pt: 34-35 °C (1.013 hPa)
Melting Pt: -116 °C (1.013 hPa)
Density: 0.71 g/cm³ (20 °C)
Flash Pt: -45 °C (1.013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00011646
CAS Number: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=9e4ec8d25df2fecee88827e09071c0f955486274&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 99.0 %
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.03 %
Titrable Acid Max. 0.0002 meq/g
Peroxides (as H₂O₂) Max. 1 ppm
Ethanol (GC) Max. 0.05 %
Carbonyl Compounds (as HCHO) Max. 0.001 %
Assay BHT (GC) Min. 1 ppm
APHA Max. 10
Vapor Pressure 8.56 psi ( 20 °C)
Refractive Index n20/D 1.3530 (lit.)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR