Isoamyl alcohol ≥98.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Warning

Synonyms: Isopentyl alcohol, iso-Pentanol, 3-Methyl-1-butanol, 3-Methylbutanol

59090-1L 59090-200L
BJ59090-1LEA 210.14 USD
BJ59090-1L BJ59090-200L
Isoamyl alcohol ≥98.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Isoamyl alcohol

ACS GENERAL USE SOLVENTS


  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications


Beilstein Number: 1718835
RTECS Number: EL5425000
UNSPSC Code: 12190000

Formula: (CH₃)₂CHCH₂CH₂OH
MW: 88.15 g/mol
Boiling Pt: 131-132 °C
Melting Pt: -117 °C
Density: 0.81 g/cm³ (20 °C)
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00002934
CAS Number: 123-51-3
EINECS: 204-633-5
UN: 1105
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd7b89cc1890b5d4b2083195c30c0d1047c32e9a&vid=170c60e26c7b4edab6e0686f91d04e676da16e6d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 98.5 %
Refractive index (n 20/D) 1.406 - 1.408
Non-volatile matter Max. 0.003 %
Water (Karl Fischer) Max. 0.3 %
Free acid (as CH₃COOH) Max. 0.01 %
Acids, esters (as amyl acetate) Max. 0.06 %
Titrable Acid Max. 0.002 meq/g
Aluminium (Al) Max. 0.5 ppm
Barium (Ba) Max. 0.1 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.5 ppm
Lithium (Li) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.02 ppm
Molybdenum (Mo) Max. 0.1 ppm
Sodium (Na) Max. 2 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.1 ppm
Strontium (Sr) Max. 0.1 ppm
Zinc (Zn) Max. 0.1 ppm
2-methylbutanol (GC) Max. 0.1 %
Carbonyl Compounds (as HCHO) Max. 0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR