Diethyl ether, anhydrous ≥99.0% contains inhibitor ACS, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
Stabilization:
Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
346136-1L
346136-6X1L
BJ346136-1LEA
197
USD
BJ346136-1L
BJ346136-6X1L
Diethyl ether, anhydrous ≥99.0% contains inhibitor ACS, Riedel-de Haën™
Diethyl ether
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Beilstein Number: 1696894
Formula:
(CH₃CH₂)₂O MW: 74.12 g/mol Boiling Pt: 34…35 °C (1.013 hPa) Melting Pt: –116 °C (1.013 hPa) Density: 0.71 g/cm³ (20 °C) Flash Pt: –45 °C (1.013 hPa) Storage Temperature: Ambient |
MDL Number:
MFCD00011646 CAS Number: 60-29-7 EINECS: 200-467-2 UN: 1155 ADR: 3,I |
Specification Test Results
IR spectrum | complying |
Assay (GC) | Min. 99.0 % |
Non-volatile matter | Max. 0.001 % |
Water (Karl Fischer) | Max. 0.03 % |
Titrable Acid | Max. 0.0002 meq/g |
Carbonyl compounds (as CO) | Max. 0.001 % |
Ethanol (GC) | Max. 0.02 % |
Methanol (GC) | Max. 0.02 % |
Peroxides (as H₂O₂) | Max. 1 ppm |
APHA | Max. 10 |
Assay BHT (GC) | Min. 1 ppm |
Refractive Index | n20/D 1.3530 (lit.) |
Explosion Limit | 36.5 % |
Contains | BHT as inhibitor |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...