Sodium silicate solution, Reagent Grade

Supplier: Honeywell Research Chemicals
Danger

338443-1L 338443-3L
BJ338443-1LEA 75 USD
BJ338443-1L BJ338443-3L
Sodium silicate solution, Reagent Grade
Sodium silicate solutions
Formula: Na₂O(SiO₂)x·xH₂O
Boiling Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003492
CAS Number: 1344-09-8
ADR: 8,III

Order Now

Specification Test Results

Assay of NaOH 13.4 - 14.4 %
Assay of SiO₂ 25.7 - 27.8 %
Composition SiO₂, ~26.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR