N,N-Dimethylformamide ≥99.8% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Research Chemicals
|
Danger
|
33120-2.5L-PE
33120-25L
33120-1L
33120-2.5L
33120-4X2.5L
33120-6X1L
BJ33120-2.5LPEEA
326.29
USD
BJ33120-2.5LPE
BJ33120-25L
BJ33120-180KG
BJ33120-1L
BJ33120-2.5L
BJ33120-4X2.5L
BJ33120-6X1L
N,N-Dimethylformamide ≥99.8% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
N,N-Dimethylformamide
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Beilstein Number: 605365
RTECS Number: LQ2100000
UNSPSC Code: 12191602
Formula:
HCON(CH₃)₂ Boiling Pt: 152-153 °C (1.013 hPa) Melting Pt: -61 °C Density: 0.94 g/cm³ (20 °C) Flash Pt: 57.5 °C (1.013.25 hPa) Storage Temperature: Ambient |
MDL Number:
MFCD00003284 CAS Number: 68-12-2 EINECS: 200-679-5 UN: 2265 ADR: 3,III |
|
Specification Test Results
Assay (GC) | Min. 99.8 % |
Density (D 20/4) | 0.948 - 0.950 |
Refractive index (n 20/D) | 1.4290 - 1.4310 |
Non-volatile matter | Max. 0.001 % |
Water (Karl Fischer) | Max. 0.1 % |
Titrable Base | Max. 0.003 meq/g |
Titrable Acid | Max. 0.0005 meq/g |
Aluminium (Al) | Max. 0.00005 % |
Boron (B) | Max. 0.000002 % |
Barium (Ba) | Max. 0.00001 % |
Bismuth (Bi) | Max. 0.00001 % |
Calcium (Ca) | Max. 0.00005 % |
Cadmium (Cd) | Max. 0.000005 % |
Cobalt (Co) | Max. 0.000002 % |
Chromium (Cr) | Max. 0.000002 % |
Copper (Cu) | Max. 0.000002 % |
Iron (Fe) | Max. 0.00001 % |
Potassium (K) | Max. 0.00005 % |
Lithium (Li) | Max. 0.00001 % |
Magnesium (Mg) | Max. 0.00001 % |
Manganese (Mn) | Max. 0.000002 % |
Molybdenum (Mo) | Max. 0.00001 % |
Sodium (Na) | Max. 0.0001 % |
Nickel (Ni) | Max. 0.000002 % |
Lead (Pb) | Max. 0.00001 % |
Tin (Sn) | Max. 0.00001 % |
Strontium (Sr) | Max. 0.00001 % |
Zinc (Zn) | Max. 0.00001 % |
APHA | Max. 15 |
Refractive Index | n20/D 1.4290-1.4310 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...