Cyclohexane ≥99.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

33117-1L 33117-2.5L 33117-4X2.5L 33117-6X1L
BJ33117-30KGEA 750.71 USD
BJ33117-30KG BJ33117-1L BJ33117-2.5L BJ33117-4X2.5L BJ33117-5L BJ33117-6X1L
Cyclohexane ≥99.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Cyclohexane

ACS GENERAL USE SOLVENTS


  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications


Beilstein Number: 1900225
RTECS Number: GU6300000
UNSPSC Code: 12191504

Formula: C₆H₁₂
Boiling Pt: 81 °C (1.013 hPa)
Melting Pt: 6.4 °C
Density: 0.78 g/cm³ (20 °C)
Flash Pt: 8.3 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00003814
CAS Number: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance complying
Assay (GC) Min. 99.5 %
Solidification range 5.5 - 6.5 °C
Boiling range 79 - 81 °C
Vapor Density 2.9 (vs air)
Transition Temp solidification point 5.5-6.5 °C
Density (D 20/4) 0.778 - 0.779
Refractive index (n 20/D) 1.4250 - 1.4280
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.01 %
Free acid (as HCl) Max. 0.001 %
Aluminium (Al) Max. 0.00005 %
Boron (B) Max. 0.000002 %
Barium (Ba) Max. 0.00001 %
Calcium (Ca) Max. 0.00005 %
Cadmium (Cd) Max. 0.000005 %
Cobalt (Co) Max. 0.000002 %
Chromium (Cr) Max. 0.000002 %
Copper (Cu) Max. 0.000002 %
Iron (Fe) Max. 0.00001 %
Magnesium (Mg) Max. 0.00001 %
Manganese (Mn) Max. 0.000002 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.00001 %
Tin (Sn) Max. 0.00001 %
Zinc (Zn) Max. 0.00001 %
Aromatic substances (as C₆H₆) Max. 0.05 %
cyclohexene (C₆H₁₀) Max. 0.05 %
Reaction against H₂SO₄ complying
APHA Max. 10


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR