1-Butanol ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Research Chemicals
|
Danger
|
33065-30KG
33065-1L
33065-2.5L
33065-4X2.5L
33065-6X1L
BJ33065-30KGEA
1013.69
USD
BJ33065-30KG
BJ33065-1L
BJ33065-2.5L
BJ33065-4X2.5L
BJ33065-6X1L
1-Butanol ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
1-Butanol
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Beilstein Number: 969148
Formula:
H₃C(CH₂)₂CH₂OH Boiling Pt: 119 °C (1.013 hPa) Melting Pt: <-90 °C Density: 0.81 g/cm³ (20 °C) Flash Pt: 9.4 %(V) Storage Temperature: Ambient |
MDL Number:
MFCD00002964 CAS Number: 71-36-3 UN: 1120 ADR: 3,III |
|
Specification Test Results
Assay (GC) | Min. 99.5 % |
Boiling range | 116 - 118 °C |
Refractive Index | n20/D 1.399 (lit.) |
Density (D 20/20) | 0.808 - 0.812 |
Non-volatile matter | Max. 0.001 % |
Water (Karl Fischer) | Max. 0.1 % |
Free acid (as C₃H₇COOH) | Max. 0.005 % |
Aluminium (Al) | Max. 0.00005 % |
Boron (B) | Max. 0.000002 % |
Barium (Ba) | Max. 0.00001 % |
Calcium (Ca) | Max. 0.00005 % |
Cadmium (Cd) | Max. 0.000005 % |
Cobalt (Co) | Max. 0.000002 % |
Chromium (Cr) | Max. 0.000002 % |
Copper (Cu) | Max. 0.000002 % |
Iron (Fe) | Max. 0.00001 % |
Magnesium (Mg) | Max. 0.00001 % |
Manganese (Mn) | Max. 0.000002 % |
Nickel (Ni) | Max. 0.000002 % |
Lead (Pb) | Max. 0.00001 % |
Tin (Sn) | Max. 0.00001 % |
Zinc (Zn) | Max. 0.00001 % |
iso-butanol (GC) | Max. 0.15 % |
n-Butyraldehyde (GC) | Max. 0.01 % |
Di-n-butyl ether (GC) | Max. 0.1 % |
2-Butanol (GC) | Max. 0.05 % |
Carbonyl compounds (as CO) | Max. 0.01 % |
APHA | Max. 10 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...