Nickel(II) chloride hexahydrate ≥98%, Puriss. p.a., Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Nickel dichloride hexahydrate

31462-1KG 31462-2.5KG
BJ31462-1KGEA 327.52 USD
BJ31462-1KG BJ31462-2.5KG
Nickel(II) chloride hexahydrate ≥98%, Puriss. p.a., Fluka™
Nickel(II) chloride hexahydrate
Formula: NiCl₂·6H₂O
MW: 237.69 g/mol
Melting Pt: 80 °C
Density: 3.550 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149809
CAS Number: 7791-20-0
UN: 3288
ADR: 6.1,III

Order Now

Specification Test Results

Assay Min. 98.0 %
pH (5 %, 20°C) 4-7
Calcium (Ca) Max. 0.005 %
Cadmium (Cd) Max. 0.001 %
Cobalt (Co) Max. 0.01 %
Copper (Cu) Max. 0.001 %
Iron (Fe) Max. 0.002 %
Magnesium (Mg) Max. 0.001 %
Sodium (Na) Max. 0.005 %
Lead (Pb) Max. 0.001 %
Zinc (Zn) Max. 0.001 %
Sulfate (SO₄) Max. 0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR