Trifluoroacetic acid ≥99%, BioSyn™ for protein sequencing, purified by redistillation, Fluka™

Supplier: Honeywell Research Chemicals
Danger

299537-25G 299537-500G 299537-100G
BJ299537-25GEA 109 USD
BJ299537-25G BJ299537-500G BJ299537-100G
Trifluoroacetic acid ≥99%, BioSyn™ for protein sequencing, purified by redistillation, Fluka™
Trifluoroacetic acid
Formula: CF₃COOH
MW: 114.02 g/mol
Boiling Pt: 72 °C (1.013 hPa)
Melting Pt: –15 °C
Density: 1.48 g/cm³ (20 °C)
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00004169
CAS Number: 76-05-1
EINECS: 200-929-3
UN: 2699
ADR: 8,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 99.0 %
Assay 99.0 - 101.0 %
Non-volatile matter Max. 0.005 %
Water (Karl Fischer) Max. 0.05 %
Suitability complying
APHA Max. 25

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR