Petroleum spirit, 100-140 °C, Puriss.

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

24534-1L 24534-25L 24534-2.5L
BJ24534-1LEA 113.07 USD
BJ24534-1L BJ24534-25L BJ24534-2.5L
Petroleum spirit, 100-140 °C, Puriss.
Petroleum spirit

We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:


  • Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
  • Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.


RTECS Number: OI6180000
UNSPSC Code: 12191502

Boiling Pt: 90-165 °C (1.013 hPa)
Density: 0.725…0.748 g/cm³ (20 °C)
Flash Pt: <10 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 920-750-0
UN: 3295
ADR: 3,II

Order Now

Specification Test Results

Boiling range 100 - 140 °C
Density (D 20/4) 0.720 - 0.745
Refractive index (n 20/D) 1.403 - 1.412
Non-volatile matter Max. 0.005 %
Water (Karl Fischer) Max. 0.01 %
Aromatic substances (as C₆H₅CH₃) Max. 0.02 %
S-compounds (as S) Max. 0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR