Tin(II) chloride dihydrate 98% ACS, Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Stannous chloride dihydrate

243523-12KG 243523-2.5KG
BJ243523-12KGEA 2784.39 USD
BJ243523-12KG BJ243523-2.5KG
Tin(II) chloride dihydrate 98% ACS, Fluka™
Tin(II) chloride dihydrate
Formula: SnCl₂·2H₂O
MW: 225.65 g/mol
Boiling Pt: Dec. (on heating)
Melting Pt: 37…38 °C
Density: 2.71 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149863
CAS Number: 10025-69-1
UN: 3260
ADR: 8,III

Order Now

Specification Test Results

Assay 98.0 - 103.0 %
Solubility in HCl complying
Calcium (Ca) Max. 0.005 %
Iron (Fe) Max. 0.003 %
Potassium (K) Max. 0.005 %
Sodium (Na) Max. 0.01 %
Lead (Pb) Max. 0.01 %
Sulfate (SO₄) Max. 0.003 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR