Acetone ≥99% (by GC), Puriss., meets analytical specification of BP, Ph. Eur., NF, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
|
Danger
|
24201-155KG
24201-20KG
24201-25L
24201-1L
24201-2.5L
24201-4X2.5L
24201-4X5L
24201-5L
24201-6X1L
24201-2.5L-GL
24201-4X2.5L-GL
BJ24201-10LEA
253.12
USD
BJ24201-10L
BJ24201-155KG
BJ24201-20KG
BJ24201-25L
BJ24201-1L
BJ24201-2.5L
BJ24201-4X2.5L
BJ24201-4X5L
BJ24201-5L
BJ24201-6X1L
BJ242012.5LGL
BJ2420142.5LGL
Acetone ≥99% (by GC), Puriss., meets analytical specification of BP, Ph. Eur., NF, Riedel-de Haën™
Acetone
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Beilstein Number: 635680
Formula:
CH₃COCH₃ Boiling Pt: 55…57 °C (1.013 hPa) Melting Pt: –95 °C Density: 0.79 g/cm³ (20 °C) Flash Pt: –17 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008765 CAS Number: 67-64-1 EINECS: 200-662-2 UN: 1090 ADR: 3,II |
|
Specification Test Results
identity (IR) | complying |
assay (GC) | Min. 99.0 % |
density (D 20/4) | 0.790 - 0.792 |
density (D 25/25) | Max. 0.789 |
non-volatile matter | Max. 20 ppm |
water (Karl Fischer) | Max. 0.3 % |
acidity or alkalinity | complying |
heavy metals (as Pb) | Max. 0.0001 % |
related subst. (GC) | complying |
benzene (GC) | Max. 2 Vol.ppm |
methanol (GC) | Max. 0.05 Vol.% |
2-propanol (GC) | Max. 0.05 Vol.% |
reducing impurities | complying |
water insoluble substances | complying |
residual solvents | complying |
appearance of the solution | complying |
Vapor Pressure | 184 mmHg ( 20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...