2-Propanol ≥99.5% (by GC), Puriss., meets analytical specification of BP, Ph. Eur., USP, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol

24137-20KG 24137-25L 24137-1L 24137-2.5L 24137-4X2.5L 24137-4X5L 24137-5L 24137-6X1L
BJ24137-160KGEA 2379.02 USD
BJ24137-160KG BJ24137-20KG BJ24137-25L BJ24137-1L BJ24137-2.5L BJ24137-4X2.5L BJ24137-4X5L BJ24137-5L BJ24137-6X1L
2-Propanol ≥99.5% (by GC), Puriss., meets analytical specification of BP, Ph. Eur., USP, Riedel-de Haën™
2-Propanol

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.


Beilstein Number: 635639

Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82
Melting Pt: -89
Density: 0.784…0.786 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3 II
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance complying
Identity (IR) complying
Assay (GC) Min. 99.5 %
Density (D 20/20) 0.785 - 0.788
Density (D 25/25) 0.783 - 0.787
Refractive index (n 20/D) 1.377 - 1.378
Non-volatile matter Max. 20 ppm
Water (Karl Fischer) Max. 0.2 %
Acidity or alkalinity complying
Acidity complying
Iron (Fe) Max. 0.0001 %
Heavy metals (as Pb) Max. 0.0002 %
Aldehydes, Ketones (as CH₃CocH₃) Max. 0.05 %
Benzene Max. 0.0001 %
Benzene and related substances complying
Methanol (GC) Max. 0.02 %
Peroxides complying
Absorbance complying
Volatile impurities (GC) complying
Residual solvents complying
Autoignition Temperature 750 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR