Toluene ≥99.3%, Laboratory Reagent

Supplier: Honeywell Research Chemicals
Danger

179965-10L 179965-2.5L 179965-1L 179965-6X1L
BJ179965-10LEA 416.55 USD
BJ179965-10L BJ179965-2.5L BJ179965-1L BJ179965-6X1L
Toluene ≥99.3%, Laboratory Reagent
Toluene

  • ≥95% for general laboratory use.


Beilstein Number: 635760

Formula: C₆H₅CH₃
Boiling Pt: 110.6 °C
Melting Pt: –95 °C
Density: 0.867 g/cm³ (20 °C)
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 99.3 %
Non-volatile matter Max. 0.005 %
Water (Karl Fischer) Max. 0.05 %
Refractive Index n20/D 1.496 (lit.)
Autoignition Temperature 997 °F
Vapor Pressure 22 mmHg ( 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR