Methanol ≥99.8% ACS, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
|
Danger
|
179337-4X4L
179337-6X1L
179337-500ML
179337-6X500ML
BJ179337-4LCS
696.53
USD
BJ179337-4L
BJ179337-1L
BJ179337-20L
BJ179337-500ML
BJ1793376500ML
Methanol ≥99.8% ACS, Riedel-de Haën™
Methanol
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Beilstein Number: 1098229
RTECS Number: PC1400000
UNSPSC Code: 12191601
Formula:
CH₃OH Boiling Pt: 64.7 °C Melting Pt: –98 °C Density: 0.792 g/cm³ (20 °C) Flash Pt: 11 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004595 CAS Number: 67-56-1 EINECS: 200-659-6 UN: 1230 ADR: 3,II |
|
Specification Test Results
Identity (IR) | complying |
Assay (GC) | min. 99.80 % |
Non-volatile matter | max. 0.001 % |
Water (Karl Fischer) | max. 0.1 % |
Titrable Acid | max. 0.0003 meq/g |
Titrable Base | max. 0.0002 meq/g |
KMnO4 reducing matter | complying |
Acetone (GC) | max. 0.001 % |
Formaldehyde | max. 0.001 % |
Acetaldehyde | max. 0.001 % |
Mixable with H2O | complying |
Reaction against H2SO4 | complying |
APHA | max. 10 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...