Cyclohexane ≥99% ACS, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
|
Danger
|
179191-1L
179191-6X1L
BJ179191-1LEA
160.98
USD
BJ179191-1L
BJ179191-6X1L
Cyclohexane ≥99% ACS, Riedel-de Haën™
Cyclohexane
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Beilstein Number: 1900225
RTECS Number: GU6300000
UNSPSC Code: 12191504
Formula:
C₆H₁₂ Boiling Pt: 81 °C (1.013 hPa) Melting Pt: 6.4 °C Density: 0.78 g/cm³ (20 °C) Flash Pt: -18 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003814 CAS Number: 110-82-7 EINECS: 203-806-2 UN: 1145 ADR: 3,II |
|
Specification Test Results
Vapor Pressure | 77 mmHg ( 20 °C) |
IR spectrum | complying |
Assay (GC) | Min. 99.0 % |
Non-volatile matter | Max. 0.002 % |
Water (Karl Fischer) | Max. 0.02 % |
Reaction against H₂SO₄ | complying |
UV-absorption | complying |
APHA | Max. 10 |
Refractive Index | n20/D 1.426 (lit.) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...